VISOCOLOR® Colorimetric Tests, Chlorine
10, VISOCOLOR® ECO Chlorine 6, refill pack, test no. 5-17;Measuring range on PF-12Plus. Range on other photometers can be different. Only for photometric evaluation, Reagents for 200 tests in a box, 0,05 - 6,00 mg/L (Cl₂); 0,05 - 6,00 mg/L (Cl₂), DPD, PF-12Plus; PF-3 Fish; PF-3 Pool; PF-3 Drinking Water; Advance, 170×115×70 mm (Including packaging), Sea water compatible; Hazardous material: No, 200, 1 KIT
Supplier: MACHEREY-NAGEL
VISOCOLOR® Colorimetric Tests, Chlorine
Catalog # MANA931217
Supplier: MACHEREY-NAGEL
CAS Number:
Jump to:
Product Details & Documents
Reagents for the determination of chlorine in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
IFU Colorimetric Test Kit VISOCOLOR Alpha Chlorine.pdf IFU Colorimetric Test Kit VISOCOLOR ECO Chlorine 1 Free and Total.pdf IFU Colorimetric Test Kit VISOCOLOR ECO Chlorine 2.pdf IFU Colorimetric Test Kit VISOCOLOR ECO Chlorine 6.pdf IFU Colorimetric Test Kit VISOCOLOR ECO Swimming pool Chlorine pH.pdf
More
Specifications
Specifications
- Chemická metoda:DPD
- Bal.:1 KIT
- Popis:VISOCOLOR® ECO Chlorine 6, refill pack, test no. 5-17;Measuring range on PF-12Plus. Range on other photometers can be different. Only for photometric evaluation
- Rozměry:170×115×70 mm (Including packaging)
- Sekvence:10
- Typ:Sea water compatible; Hazardous material: No
- Pro jednotku:PF-12Plus; PF-3 Fish; PF-3 Pool; PF-3 Drinking Water; Advance
- Testů v kitu:200
- Rozsah:0,05 - 6,00 mg/L (Cl₂); 0,05 - 6,00 mg/L (Cl₂)
- Obsahy:Reagents for 200 tests in a box
- Hmotnost:147,45 g (Including packaging)
- Cat. No.:MANA931217
- Plošina:Photometric test kits
- Životnost police (měsíce):24
- Model:MANA931217
- Skladovací teplota:15...25 °C



