Cryoboxes
02, 4×4, 50 ml tubes, —, - mm
Katalogové číslo HEATHS2860P
Dodavatel: HEATHROW
Přejít na:
Product Details & Documents
Made of cardboard, white.
- Write-on surface for descriptions or self numbering systems
- Dividers are flat packed and comes in two different sizes (to be ordered separately)
For storage of large samples and 15 or 50 ml tubes.
Documents
HEATHS2860M_TDS_EN
Technický list
Specifikace
- Přihrádky:4×4
- Určeno pro:50 ml tubes
- Š×H×V vnějš.:- mm
- Barva:—
- Sekvence:02