Orders LinkContactUsLinkComponent
Separating funnels, conical
Separating funnels, conical
250 ml, 29/32 NS
Katalogové číslo 532-0105
Dodavatel:   Lenz Laborglas
Separating funnels, conical
Katalogové číslo 532-0105
Dodavatel:  Lenz Laborglas
Separating funnels, conical
Separating funnels, conical
250 ml, 29/32 NS
Přejít na:
Price
6 770,00 Kč
Přihlásit se pro dostupnost
1 * 2 KS
Product Details & Documents
Separating funnels, conical, made of borosilicate glass 3.3, amber coloured.
  • Amber coloured to protect light sensitive substances
  • Large labeling field
  • Very good chemical and temperature resistance
With interchangeable PTFE key.
Documents
Specifikace
  • Velikost objímky:
    29/32 NS
  • Kapacita:
    250 ml