Orders LinkContactUsLinkComponent
Test panels
Test panels
10, Test panel, aluminium, chessboard printed, degreased and cleaned, 100×300×0,5 mm
Katalogové číslo SIMEVF8518
Dodavatel:   Industrial Physics
Test panels
Katalogové číslo SIMEVF8518
Dodavatel:  Industrial Physics
Test panels
Test panels
10, Test panel, aluminium, chessboard printed, degreased and cleaned, 100×300×0,5 mm
Přejít na:
Price
69 430,00 Kč
Přihlásit se pro dostupnost
1 * 300 KS
Product Details & Documents
Test panels are used to enhance reproducibility of physical and chemical tests. Each panel is equipped with a hole for hanging and handling.
  • Consistent and reproducible quality with sophisticated finishing: No burrs or sharp edges
  • Physical testing: Adhesion, elongation, impact resistance and elasticity
  • Appearance testing of coatings (gloss, colour, texture)
Documents
Specifikace
  • Popis:
    Test panel, aluminium, chessboard printed, degreased and cleaned
  • Sekvence:
    10
  • Š×H×V:
    100×300×0,5 mm