Orders LinkContactUsLinkComponent
Specific gravity cups, calibrated
Specific gravity cups, calibrated
02, Anodised aluminium, 100 ml
Katalogové číslo SIMEVF2097
Dodavatel:   Industrial Physics
Specific gravity cups, calibrated
Katalogové číslo SIMEVF2097
Dodavatel:  Industrial Physics
Specific gravity cups, calibrated
Specific gravity cups, calibrated
02, Anodised aluminium, 100 ml
Přejít na:
Price
9 030,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Specific gravity cups for determining the specific gravity or density (or weight per gallon wpg) of coatings, pastes or similar liquids.
  • Density is defined as weight per unit volume at a specified temperature
  • Available in anodised aluminium or stainless steel
Certifications: According to ISO 2811, DIN 53 217 and ASTM D 1475.
Informace o dodávce: Supplied with a calibration certificate.
Documents
Specifikace
  • Objem (nom.):
    100 ml
  • Popis:
    Anodised aluminium
  • Sekvence:
    02