Orders LinkContactUsLinkComponent
Tank tool
01, Reverse tank tool adapter, electropolished stainless steel
Katalogové číslo 129-1072
Dodavatel:   MICRONOVA
Tank tool
Katalogové číslo 129-1072
Dodavatel:  MICRONOVA
Tank tool
Tank tool
01, Reverse tank tool adapter, electropolished stainless steel
Přejít na:
Price
10 070,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
The tank tool is designed to efficiently clean chambers, tanks and pipes from above or below. The articulated head allows curved frame to hug the outer wall of the tank as it moves downward to remove residue or apply disinfectant. The unit has a degree of swivel to ensure good contact with the surface being cleaned.
  • Stainless steel frame adapter with swivel action
  • Curved frame maintains contact while cleaning tanks
  • Available in exterior and interior (reverse) configurations
  • Designed for cleanroom use
Frame adapters hold MZSM series of replaceable mop heads. Covers available in a variety of fabrics.
Attaches directly to SSU series handles. Attaches to STAH and STAEH series via SSA-2 Adapter.
Specifikace
  • Popis:
    Reverse tank tool adapter, electropolished stainless steel
  • Sekvence:
    01