Orders LinkContactUsLinkComponent
Accessories for Combitips® advanced
Accessories for Combitips® advanced
01, Adapter 25 ml for Combitips® Advanced
Catalog # 613-2085
   Low Manufacturing Impact|Product Transparency and Disclosure
Supplier:   EPPENDORF
Accessories for Combitips® advanced
Catalog # 613-2085
Supplier:  EPPENDORF
   Low Manufacturing Impact|Product Transparency and Disclosure
Accessories for Combitips® advanced
Accessories for Combitips® advanced
01, Adapter 25 ml for Combitips® Advanced
Price
308,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Specifications
  • Popis:
    Adapter 25 ml for Combitips® Advanced
  • Sekvence:
    01
  • Certifikát ochrany životního prostředí:
    Renewable Energy Certificate, ISO 14001, ACT Label
  • Environmentally Preferable:
    Low Manufacturing Impact|Product Transparency and Disclosure
  • Cat. No.:
    613-2085