Orders LinkContactUsLinkComponent
Accessories for Combitips® advanced
Accessories for Combitips® advanced
03, Adapter 50 ml for Combitips® Advanced
Katalogové číslo 613-2086
   Low Manufacturing Impact|Product Transparency and Disclosure
Dodavatel:   EPPENDORF
Accessories for Combitips® advanced
Katalogové číslo 613-2086
Dodavatel:  EPPENDORF
   Low Manufacturing Impact|Product Transparency and Disclosure
Accessories for Combitips® advanced
Accessories for Combitips® advanced
03, Adapter 50 ml for Combitips® Advanced
Přejít na:
Price
308,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Specifikace
  • Popis:
    Adapter 50 ml for Combitips® Advanced
  • Sekvence:
    03
  • Cat. No.:
    613-2086
  • Certifikát ochrany životního prostředí:
    Renewable Energy Certificate, ISO 14001, ACT Label
  • Environmentally Preferable:
    Low Manufacturing Impact|Product Transparency and Disclosure