Orders LinkContactUsLinkComponent
Accessories for Combitips® advanced
02, Adapter 25 ml for Combitips® Advanced Biopur®
  613-2087
   Low Manufacturing Impact|Product Transparency and Disclosure
 :   EPPENDORF
Accessories for Combitips® advanced
  613-2087
 :  EPPENDORF
   Low Manufacturing Impact|Product Transparency and Disclosure
Accessories for Combitips® advanced
Accessories for Combitips® advanced
02, Adapter 25 ml for Combitips® Advanced Biopur®
2 250,00 Kč
1 * 7 KS
  • Popis:
    Adapter 25 ml for Combitips® Advanced Biopur®
  • Sekvence:
    02
  • Cat. No.:
    613-2087
  • Certifikát ochrany životního prostředí:
    Renewable Energy Certificate, ISO 14001, ACT Label
  • Environmentally Preferable:
    Low Manufacturing Impact|Product Transparency and Disclosure