Orders LinkContactUsLinkComponent
Accessories for Combitips® advanced
Accessories for Combitips® advanced
04, Adapter 50 ml for Combitips® Advanced Biopur®
Katalogové číslo 613-2088
   Low Manufacturing Impact|Product Transparency and Disclosure
Dodavatel:   EPPENDORF
Accessories for Combitips® advanced
Katalogové číslo 613-2088
Dodavatel:  EPPENDORF
   Low Manufacturing Impact|Product Transparency and Disclosure
Accessories for Combitips® advanced
Accessories for Combitips® advanced
04, Adapter 50 ml for Combitips® Advanced Biopur®
Přejít na:
Price
2 250,00 Kč
Přihlásit se pro dostupnost
1 * 7 KS
Product Details & Documents
Specifikace
  • Popis:
    Adapter 50 ml for Combitips® Advanced Biopur®
  • Sekvence:
    04
  • Cat. No.:
    613-2088
  • Certifikát ochrany životního prostředí:
    Renewable Energy Certificate, ISO 14001, ACT Label
  • Environmentally Preferable:
    Low Manufacturing Impact|Product Transparency and Disclosure