Tanks, rectangular, LDPE, LaboPlast®
09, 115 L, Seamless containers, rectangular shaped, 500×400×600 mm
BURK3440-1156
: BURKLE
These seamless rectangular tanks feature optimum ratio of wall thickness to filling load.
- Linear LDPE, transparent
- Rupture-proof
- Kapacita:115 L
- Popis:Seamless containers, rectangular shaped
- Sekvence:09
- Š×H×V:500×400×600 mm