Orders LinkContactUsLinkComponent
Tanks, rectangular, LDPE, LaboPlast®
Tanks, rectangular, LDPE, LaboPlast®
16, 260 L, Seamless containers, rectangular shaped, 800×600×600 mm
Katalogové číslo BURK3440-2607
Dodavatel:   BURKLE
Tanks, rectangular, LDPE, LaboPlast®
Katalogové číslo BURK3440-2607
Dodavatel:  BURKLE
Tanks, rectangular, LDPE, LaboPlast®
Tanks, rectangular, LDPE, LaboPlast®
16, 260 L, Seamless containers, rectangular shaped, 800×600×600 mm
Price
20 430,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
These seamless rectangular tanks feature optimum ratio of wall thickness to filling load.
  • Linear LDPE, transparent
  • Rupture-proof
Specifikace
  • Kapacita:
    260 L
  • Popis:
    Seamless containers, rectangular shaped
  • Sekvence:
    16
  • Š×H×V:
    800×600×600 mm
Accessories
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Container thread ¾" for cylindrical containers
    Container thread ¾" for cylindrical containers
    BURK3440-0034
    01
    Přihlásit se pro dostupnost
    1 * 1 KS - 4 360,00 Kč