Orders LinkContactUsLinkComponent
Plaster dispenser, QuickFix Detectable
Plaster dispenser, QuickFix Detectable
Refill pack with 45 detectable plasters, for use in dispenser
Katalogové číslo 118-0234
Dodavatel:   Plum Safety
Plaster dispenser, QuickFix Detectable
Katalogové číslo 118-0234
Dodavatel:  Plum Safety
Plaster dispenser, QuickFix Detectable
Plaster dispenser, QuickFix Detectable
Refill pack with 45 detectable plasters, for use in dispenser
Přejít na:
Price
2 090,00 Kč
Přihlásit se pro dostupnost
1 * 6 KS
Product Details & Documents
Dispenser containing highly visible blue plasters with an integrated metal surface that can be detected via a metal detector. The dispenser contains 90 plasters (2 refills) and can be wall-mounted.
  • Elastic textile fabric: breathable and skin friendly
  • Adapt to movements of the skin and are therefore very comfortable to wear
  • Very hygienic as every plaster is individually wrapped
  • Dispenser can be operated with just one hand
  • Easy to refill
Plaster size: 72×25 mm
 : Dispenser includes 2×45 plaster strips. A refill pack can be ordered separately.
Specifikace
  • Popis:
    Refill pack with 45 detectable plasters, for use in dispenser