Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
 

Fleece jacket, DYNAMIC® Tortolas 443

06, Tmavomodrá, 3XL
Catalog # SIOE443Z/B95/3XL
Supplier:   SIOEN
Fleece jacket, DYNAMIC® Tortolas 443
Catalog # SIOE443Z/B95/3XL
Supplier:  SIOEN
Fleece jacket, DYNAMIC® Tortolas 443
Fleece jacket, DYNAMIC® Tortolas 443

06, Tmavomodrá, 3XL

Price
2 590,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents

Single-coloured jacket for men, made of 2-layer bonded polyester fleece. Both sides are brushed, one side features an anti-pilling treatment. Can be zipped into different jackets.

  • Straight collar, hidden zip closure
  • Inset chest pocket with zip closure, two inset pockets with zip closure
  • Round inset sleeves, elastic piping at end of sleeves
  • Drawstring in hem

Weight: 320 g/m²

 : EN 340 (1XXXX)

 :

 :

 :

 :

Specifications
  • Velikost oblečení:
    3XL
  • Barva:
    Tmavomodrá
  • Sekvence:
    06
  • Hmotnost:
    320 g/m²
  • Cat. No.:
    SIOE443Z/B95/3XL
  • Typ uzávěru:
    Se zipem
  • Sleeve or leg length:
    Long
  • Risk area:
    Cold
  • Tkanina:
    Polyester
  • Rod:
    Pro muže
  • Model:
    Tortolas 443
  • Aplikace oblečení:
    General Purpose
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.