Methyl palmitate
3047, Glass bottle for solids, 1 g
Dodavatel: Cayman Chemical
Přejít na:
Product Details & Documents
Synonyma: Hexadecylic acid methyl ester, Methyl hexadecanoate, Palmitic acid methyl ester, Hexadecanoic acid methyl ester, Palmitic Acid Methyl Ester
- Číslo CAS:112-39-0
- Vzorec:C₁₇H₃₄O₂
- MDL číslo:MFCD00008994
- Molecular Weight:270.46
- Bod varu:417 °C (1013 hPa)
- Bod tání:29,9 °C
- Hustota:0,8804 g/cm³ (20 °C)
- :Density: 0,8804 g/cm³ (20 °C)
- EINECS:203-966-3
Certifikáty
Hledat podle čísla šaržeSpecifikace
Specifikace
- Typ balení:Glass bottle for solids
- Bal.:1 g
- Sekvence:3047
Specifikace
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



