Orders LinkContactUsLinkComponent
Separating funnels, Squibb
500 ml, 29/32 NS
Catalog # LENZ04018058
Supplier:   Lenz Laborglas
Separating funnels, Squibb
Catalog # LENZ04018058
Supplier:  Lenz Laborglas
Separating funnels, Squibb
Separating funnels, Squibb
500 ml, 29/32 NS
Price
6 860,00 Kč
Log in for availability
1 * 2 KS
Product Details & Documents
Separating funnels, Squibb, stopcock with glass or PTFE key, made of borosilicate glass amberized.
  • Without graduation
  • Very good temperature and chemical resistance
  • Amber coloured for ligth sensitve liquids
Specifications
  • Velikost objímky:
    29/32 NS
  • Kapacita:
    500 ml