Orders LinkContactUsLinkComponent
Shakers
Shakers
Shaker for six Mojonnier tubes
Katalogové číslo 614-0184
Dodavatel:   GERBER FUNKE
Shakers
Katalogové číslo 614-0184
Dodavatel:  GERBER FUNKE
Shakers
Shakers
Shaker for six Mojonnier tubes
Price
55 190,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Shakers for determining the fat content in milk according to Mojonnier's method. White PVC housing. Choice of models to accept 4, 6 or 36 Mojonnier extraction tubes. Units provide forceful, uniform and reproducible mixing.
  • Timer, 0 to 15 min
  • Produce 100 shakes per minute
  • Model for 36 butyrometers is supplied with stand
Specifications
  • Popis:
    Shaker for six Mojonnier tubes
Accessories
Product Information
  • Katalogové číslo
    Dostupnost
    Price
  • Cork stopper for extraction tubes, Mojonnier`s method
    Cork stopper for extraction tubes, Mojonnier`s method
    614-0188
    Log in for availability
    1 * 1 KS - 7,90 Kč
Product Information
  • Katalogové číslo
    Dostupnost
    Price
  • Extraction tubes, Mojonnier, with flattened bulb
    Extraction tubes, Mojonnier, with flattened bulb
    614-0187
    Log in for availability
    1 * 1 KS - 549,00 Kč
Product Information
  • Katalogové číslo
    Dostupnost
    Price
  • Extraction tubes, Mojonnier, with rounded bulb
    Extraction tubes, Mojonnier, with rounded bulb
    614-0186
    Log in for availability
    1 * 1 KS - 549,00 Kč
Product Information
  • Katalogové číslo
    Dostupnost
    Price
  • Stand, wooden, for 12 extraction tubes acc. to Mojonnier`s method
    Stand, wooden, for 12 extraction tubes acc. to Mojonnier`s method
    614-0189
    Log in for availability
    1 * 1 KS - 2 020,00 Kč