Síran amonný
4024, Glass bottle for solids, 1 kg
Katalogové číslo RC-019
Dodavatel: G-Biosciences
Přejít na:
Product Details & Documents
Synonyma: Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite
- Číslo CAS:7783-20-2
- Vzorec:(NH₄)₂SO₄
- MDL číslo:MFCD00003391
- Molecular Weight:132.14
- Merck Index:13,00559
- Bod tání:336…339 °C
- Hustota:1,78 g/cm³ (20 °C)
- :Density: 1,78 g/cm³ (20 °C)
- Teplota pro skladování:Ambient
- EINECS:231-984-1
- REACH:01-2119455044-46
Specifikace
Specifikace
- Typ balení:Glass bottle for solids
- Bal.:1 kg
- Sekvence:4024
Specifikace
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



