Orders LinkContactUsLinkComponent
Filter discs, DURAN®
Filter discs, DURAN®
102, 5, 120 mm, 8,5 mm
Katalogové číslo SCOT251620508
Dodavatel:   DWK Life Sciences
Filter discs, DURAN®
Katalogové číslo SCOT251620508
Dodavatel:  DWK Life Sciences
Filter discs, DURAN®
Filter discs, DURAN®
102, 5, 120 mm, 8,5 mm
Přejít na:
Price
6 530,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Sintered glass filters have neutral properties with unfused edge and untreated surfaces. The hard sintering process makes the material resilient.
  • Available in intermediate sizes, alternative dimensions, alternate porosities and bi-planar design
Made from clear DURAN® borosilicate glass 3,3.
Specifikace
  • Ø:
    120 mm
  • Porozita:
    5
  • Sekvence:
    102
  • Tloušťka:
    8,5 mm