Orders LinkContactUsLinkComponent
Watchmakers' magnifiers
Watchmakers' magnifiers
003, Magnifier with aspheric lens, 10×, 25 mm
Katalogové číslo ESCH1124110
Dodavatel:   ESCHENBACH OPTIK
Watchmakers' magnifiers
Katalogové číslo ESCH1124110
Dodavatel:  ESCHENBACH OPTIK
Watchmakers' magnifiers
Watchmakers' magnifiers
003, Magnifier with aspheric lens, 10×, 25 mm
Přejít na:
Price
809,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
PXM® lightweight lenses with antistatic and scratch resistant coating. Ideal as a watchmaker's magnifier or gemstone magnifier.
  • Includes vent to prevent condensation
  • Made of flame resistant plastic material
  • Colour: anthracite
Specifikace
  • Ø:
    25 mm
  • Zvětšení:
    10×
  • Popis:
    Magnifier with aspheric lens
  • Sekvence:
    003