Pipette tips
10, Standard, 0,5 - 200 µl, +, 51 mm, Žlutá, 10 racks, 96 each
Supplier: Sartorius
Jump to:
Product Details & Documents
These pipette tips are suitable for many pipettes on the market.
- Natural colour, premium grade PP, autoclavable
- Excellent seal design for accurate, leak-tight sealing
- Produced and packed in a dust-free environment under strictly maintained conditions
Ordering information: Available in bulk packaging, in autoclavable racks and in refills.
Caution: Article 613-4153 (BHIT780310) is designed for use with the Midi Plus Pipette Controller only.
Documents
Specifications
- Barevný kód:Žlutá
- Balené:10 racks, 96 each
- Sterilní:+
- Objem:0,5 - 200 µl
- Délka:51 mm
- Sekvence:10
- Druh špičky:Standard
- Cat. No.:732-1557
- Certifikát ochrany životního prostředí:Renewable energy certificate; ISO 14001
- Environmentally Preferable:Low Manufacturing Impact