Orders LinkContactUsLinkComponent
Watchmakers' magnifiers
Watchmakers' magnifiers
002, Magnifier with plano-convex lens, 7×, 25 mm
Catalog # ESCH11247
Supplier:   ESCHENBACH OPTIK
Watchmakers' magnifiers
Catalog # ESCH11247
Supplier:  ESCHENBACH OPTIK
Watchmakers' magnifiers
Watchmakers' magnifiers
002, Magnifier with plano-convex lens, 7×, 25 mm
Price
698,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
PXM® lightweight lenses with antistatic and scratch resistant coating. Ideal as a watchmaker's magnifier or gemstone magnifier.
  • Includes vent to prevent condensation
  • Made of flame resistant plastic material
  • Colour: anthracite
Specifications
  • Ø:
    25 mm
  • Zvětšení:
  • Popis:
    Magnifier with plano-convex lens
  • Sekvence:
    002