Orders LinkContactUsLinkComponent
Padlock stations
Padlock stations
02, Large padlock station, for up to 24 padlocks, Červený, 406×31×76 mm
Catalog # BRDY800121
Supplier:   Brady
Padlock stations
Catalog # BRDY800121
Supplier:  Brady
Padlock stations
Padlock stations
02, Large padlock station, for up to 24 padlocks, Červený, 406×31×76 mm
Price
1 180,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
These stations constructed of 14 gauge steel with red plastic coating are ideal for storing padlocks when they are not in use.
  • Rugged and eye-catching stations
  • Hold up to 4 or 24 padlocks
Ordering information: Padlocks supplied seperately.
Specifications
  • Barva:
    Červený
  • Popis:
    Large padlock station, for up to 24 padlocks
  • Sekvence:
    02
  • Š×H×V:
    406×31×76 mm