Orders LinkContactUsLinkComponent
Padlock control centres
Padlock control centres
02, Padlock control centre, 18 hooks, Červený, 457×50×394 mm
Catalog # BRDY800118
Supplier:   Brady
Padlock control centres
Catalog # BRDY800118
Supplier:  Brady
Padlock control centres
Padlock control centres
02, Padlock control centre, 18 hooks, Červený, 457×50×394 mm
Price
11 580,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
These padlock control centres provide safe storage and controlled access to padlocks.
  • Made from powder-coated steel
  • Each nickel coated metal hook can hold two locks
  • Two keys are provided for cabinet lock
Ordering information: Components not included. Padlocks need to be ordered separately.
Specifications
  • Barva:
    Červený
  • Popis:
    Padlock control centre, 18 hooks
  • Sekvence:
    02
  • Š×H×V:
    457×50×394 mm