Electrode Cables
14, Adapter cable E-F, for connecting electrodes with type E plug (=DIN plug) to instruments with type F socket (mini-socket)
METR6.2104.000
: METROHM
- Popis:Adapter cable E-F, for connecting electrodes with type E plug (=DIN plug) to instruments with type F socket (mini-socket)
- Sekvence:14