Orders LinkContactUsLinkComponent
Fluorescent inks, Blak-Ray®
02, Readmittance ink, RI-20, green, Textile marking, readmittance, coding, theatrical, amusement, promotional, inventory control
  UVPA96-0021-02
Fluorescent inks, Blak-Ray®
  UVPA96-0021-02
Fluorescent inks, Blak-Ray®
Fluorescent inks, Blak-Ray®
02, Readmittance ink, RI-20, green, Textile marking, readmittance, coding, theatrical, amusement, promotional, inventory control
1 070,00 Kč
1 * 1 KS
Blak-Ray® fluorescent inks are suited for use on skin, fabric and similar porous or textured surfaces.
  • Simple to use - inks come ready made
  • Won't gum up - stamp pads stay cleaner longer
  • Economical - small amounts of ink are used with stamp pads
  • All inks are formulated to fluoresce brightly under UVP's longwave ultraviolet lamps
  • Readmission type inks are designed for use at amusement parks, social gatherings and sporting events
  • Inks are non-toxic, fast-drying and non-transferable
Suitable for readmission control, inspection, coding and inventory control.
  • K použití s:
    Textile marking, readmittance, coding, theatrical, amusement, promotional, inventory control
  • Popis:
    Readmittance ink, RI-20, green
  • Sekvence:
    02