Fluorescent inks, Blak-Ray®
01, Readmittance ink, RI-10, blue, Textile marking, readmittance, coding, theatrical, amusement, promotional, inventory control
Katalogové číslo UVPA96-0021-01
Dodavatel: UVP ULTRA VIOLET PRODUCTS
Přejít na:
Product Details & Documents
Blak-Ray® fluorescent inks are suited for use on skin, fabric and similar porous or textured surfaces.
- Simple to use - inks come ready made
- Won't gum up - stamp pads stay cleaner longer
- Economical - small amounts of ink are used with stamp pads
- All inks are formulated to fluoresce brightly under UVP's longwave ultraviolet lamps
- Readmission type inks are designed for use at amusement parks, social gatherings and sporting events
- Inks are non-toxic, fast-drying and non-transferable
Suitable for readmission control, inspection, coding and inventory control.
Specifikace
- K použití s:Textile marking, readmittance, coding, theatrical, amusement, promotional, inventory control
- Popis:Readmittance ink, RI-10, blue
- Sekvence:01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.