Orders LinkContactUsLinkComponent
Brushes, FlexBrush™
Brushes, FlexBrush™
03, Adapter, hook-shaped brush, twisted zinc with plastic handle, Polyester, Plast, 292 mm
Catalog # 129-1068
   Waste Reducing
Supplier:   MICRONOVA
Brushes, FlexBrush™
Catalog # 129-1068
Supplier:  MICRONOVA
   Waste Reducing
Brushes, FlexBrush™
Brushes, FlexBrush™
03, Adapter, hook-shaped brush, twisted zinc with plastic handle, Polyester, Plast, 292 mm
Price
1 200,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
The FlexBrush™ is ideal for cleaning over and around piping and fittings. The flexibility of the head allows the user to shape the head for custom cleaning applications.
  • Bendable brush with polyester or microfibre cover
  • Acme threaded end attaches to a variety of handles for extended reach
  • Threaded handle (STA series) can be added for extended reach
  • Covers are compatible with strong disinfectants and autoclavable
Specifications
  • Materiál štětin:
    Polyester
  • Handle material:
    Plast
  • Popis:
    Adapter, hook-shaped brush, twisted zinc with plastic handle
  • Sekvence:
    03
  • Celková délka:
    292 mm
  • Cat. No.:
    129-1068
  • Environmentally Preferable:
    Waste Reducing