Orders LinkContactUsLinkComponent
Watch glasses
Watch glasses
06, Watch glass, glass, with hole in center, 200 mm
Katalogové číslo HECH92001200020
Watch glasses
Katalogové číslo HECH92001200020
Watch glasses
Watch glasses
06, Watch glass, glass, with hole in center, 200 mm
Přejít na:
Price
265,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Evaporating dishes, made of glass, with fused rim.
  • Good chemical restistance
  • Available with or without hole
Certifications: DIN 12 341
Specifikace
  • Ø:
    200 mm
  • Popis:
    Watch glass, glass, with hole in center
  • Sekvence:
    06