Orders LinkContactUsLinkComponent
Conductivity standards
Conductivity standard (500 ml), 1413 µS, 1
  JENW16156-59
Conductivity standards
  JENW16156-59
 :  
Conductivity standards
Conductivity standard (500 ml), 1413 µS, 1
 :  
2 090,00 Kč
1 * 1 KS
Standard solutions are used for calibrating conductivity cells.
  • Bal.:
    1
  • Popis:
    Conductivity standard (500 ml)
  • Vodivost:
    1413 µS