Orders LinkContactUsLinkComponent
Gram Negative lysis buffer
Gram Negative lysis buffer
03, Gram negative lysis buffer, 500 mL
Katalogové číslo 786-579
Dodavatel:   G-Biosciences
Gram Negative lysis buffer
Katalogové číslo 786-579
Dodavatel:  G-Biosciences
Číslo CAS:  
Gram Negative lysis buffer
Gram Negative lysis buffer
03, Gram negative lysis buffer, 500 mL
Přejít na:
Číslo CAS:  
Price
5 040,00 Kč
Přihlásit se pro dostupnost
1 * 500 mL
Product Details & Documents
Gram-negative lysis buffer is an extraction buffer for soluble proteins from Gram-negative bacteria. It is a proprietary improvement on the lysozyme based lysis in combination with various salts and agents, which allows extraction of soluble proteins. This buffer has been tested for use with several widely used bacteria including E.coli strains.
Specifikace
Specifikace
  • Bal.:
    500 mL
  • Popis:
    Gram negative lysis buffer
  • Sekvence:
    03