Orders LinkContactUsLinkComponent
Filter vials, SINGLE StEP™
02, Nylon, 0,2 µm, Černá
Katalogové číslo 548-1408
Dodavatel:   THOMSON INSTRUMENT
Filter vials, SINGLE StEP™
Katalogové číslo 548-1408
Dodavatel:  THOMSON INSTRUMENT
Filter vials, SINGLE StEP™
Filter vials, SINGLE StEP™
02, Nylon, 0,2 µm, Černá
Přejít na:
Price
46 370,00 Kč
Přihlásit se pro dostupnost
1 * 500 KS
Product Details & Documents
Simple replacements for HPLC vials, HPLC caps, syringes, needles, and syringe filters. Filter vials minimise steps and loss of product. They protect HPLC, GC, GC-MS and LC-MS systems against contaminants and particulates.
  • Compatible with both UHPLC and standard HPLC autosamplers
  • Colour coded pre-slit caps for easy identification of membrane and porosity
  • Low dead volume unit contains rugged PP insert with 450 µl loading capacity
Specifikace
  • Membrána:
    Nylon
  • Velikost pórů:
    0,2 µm
  • Barva:
    Černá
  • Sekvence:
    02