Orders LinkContactUsLinkComponent
Filter vials, SINGLE StEP™
Filter vials, SINGLE StEP™
13, PVDF, 0,45 µm, Žlutá
Katalogové číslo 548-1413
Dodavatel:   THOMSON INSTRUMENT
Filter vials, SINGLE StEP™
Katalogové číslo 548-1413
Dodavatel:  THOMSON INSTRUMENT
Filter vials, SINGLE StEP™
Filter vials, SINGLE StEP™
13, PVDF, 0,45 µm, Žlutá
Přejít na:
Price
18 540,00 Kč
Přihlásit se pro dostupnost
1 * 200 KS
Product Details & Documents
Simple replacements for HPLC vials, HPLC caps, syringes, needles, and syringe filters. Filter vials minimise steps and loss of product. They protect HPLC, GC, GC-MS and LC-MS systems against contaminants and particulates.
  • Compatible with both UHPLC and standard HPLC autosamplers
  • Colour coded pre-slit caps for easy identification of membrane and porosity
  • Low dead volume unit contains rugged PP insert with 450 µl loading capacity
Specifikace
  • Membrána:
    PVDF
  • Velikost pórů:
    0,45 µm
  • Barva:
    Žlutá
  • Sekvence:
    13