Orders LinkContactUsLinkComponent
Stand rods
Stand rods
02, 12 mm, 450 mm
Catalog # 241-0379
Supplier:   USBECK Laborgeräte
Stand rods
Catalog # 241-0379
Stand rods
Stand rods
02, 12 mm, 450 mm
Price
339,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Stainless steel.
  • Robust, very stable
  • Excellent chemical resistance
Specifications
  • Ø:
    12 mm
  • Délka:
    450 mm
  • Sekvence:
    02