Stopcocks for dessicator lids
019, Stopcock for dessicator lids, borosilicate glass, with NS 24/29 cone and 14,5 mm solid glass key, bore of key 2,5 mm
Katalogové číslo WITG0.246.002
Dodavatel: witeg Labortechnik
Přejít na:
Product Details & Documents
Specifikace
- Popis:Stopcock for dessicator lids, borosilicate glass, with NS 24/29 cone and 14,5 mm solid glass key, bore of key 2,5 mm
- Sekvence:019