0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
JETPEI-MACROPHAGE TRANSFECTION REAGENT 1 * 100 µl
103-01N
:
Polyplus-transfection
JETPEI-MACROPHAGE TRANSFECTION REAGENT 1 * 100 µl
103-01N
:
Polyplus-transfection
CPT_103_jetPEI-Macrophage_vD
NHPS_103_vA
PF_jetPEI-Macrophage_vC
Frequently Bought Together
Sealing film for manual application, Parafilm® M
: HEATHROW
Aceton ≥99%, TECHNICAL
: VWR Chemicals
Voda, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
VWR® NITRILE LIGHT, Rukavice, nitrilové
: VWR Collection
VWR®, Pipety Pasteurovy, High performance
: VWR Collection
Acetonitril, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® USP, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
Acetonitril ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Rukavice, nitrilové, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR®, Sérologická pipeta, standardní
: VWR Collection
VWR®, Ubrousky čisticí
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals