Orders LinkContactUsLinkComponent
Separating funnels, conical
Separating funnels, conical
500 ml, 29/32 NS
Katalogové číslo WITG2.676.500
Dodavatel:   witeg Labortechnik
Separating funnels, conical
Katalogové číslo WITG2.676.500
Dodavatel:  witeg Labortechnik
Separating funnels, conical
Separating funnels, conical
500 ml, 29/32 NS
Přejít na:
Price
5 280,00 Kč
Přihlásit se pro dostupnost
1 * 2 KS
Product Details & Documents
Separating funnels, conical, made of DURAN® borosilicate glass 3.3, graduated, with Witaflo-needle valve stopcock and PTFE-needle spindle.
  • Easy to read graduation
  • With plastic stopper
  • Very good chemical and temperature resistance
Specifikace
  • Velikost objímky:
    29/32 NS
  • Kapacita:
    500 ml