Orders LinkContactUsLinkComponent
Separating funnels, conical
1000 ml, 29/32 NS
Catalog # WITG2.676.001
Supplier:   witeg Labortechnik
Separating funnels, conical
Catalog # WITG2.676.001
Supplier:  witeg Labortechnik
Separating funnels, conical
Separating funnels, conical
1000 ml, 29/32 NS
Price
7 030,00 Kč
Log in for availability
1 * 2 KS
Product Details & Documents
Separating funnels, conical, made of DURAN® borosilicate glass 3.3, graduated, with Witaflo-needle valve stopcock and PTFE-needle spindle.
  • Easy to read graduation
  • With plastic stopper
  • Very good chemical and temperature resistance
Specifications
  • Velikost objímky:
    29/32 NS
  • Kapacita:
    1000 ml