Temperature indicating kit
01, Thermax chill checker, type 3, -5 °C
Katalogové číslo KLEI1211103
Dodavatel: KLEINFELD
Přejít na:
Product Details & Documents
These Thermax chill checker include eight different measurement points over a temperature range from −17 to +20 °C. Activation takes place after cooling by pressing in the center. When the temperature is exceeded, it changes color purple or blue.
Specifikace
- Max. tepl.:-5 °C
- Popis:Thermax chill checker, type 3
- Sekvence:01



