Pycnometers for bituminous materials
01, Pycnometer with ring mark, matt shield and numbering, soda galss, 1000 ml, 60
Katalogové číslo NEUB0321-50-1000
Dodavatel: NEUBERT VOLUME GLASSWAERE
Pycnometers for bituminous materials
Katalogové číslo NEUB0321-50-1000
Dodavatel: NEUBERT VOLUME GLASSWAERE
Přejít na:
Product Details & Documents
Soda-lime glass or borosilicate glass 3.3.
- Two-part assembly both parts with identification number
- Cap with funnel for ease of filling
- Ring mark on cap stem
Specifikace
- NZ:60
- Objem (nom.):1000 ml
- Popis:Pycnometer with ring mark, matt shield and numbering, soda galss
- Sekvence:01