Microplates, 96-well
04, 96-well microplate, F-bottom (standard), MICROLON® 200, individually packed, 25 - 340 µl, 382 µl
Katalogové číslo GREI655001-000
Dodavatel: Greiner Bio-One
Microplates, 96-well
Katalogové číslo GREI655001-000
Dodavatel: Greiner Bio-One
Přejít na:
Product Details & Documents
Crystal clear PS, without lid. Ideal for ELISA methodologies, these plates are available with high binding surfaces that display a higher protein binding capacity for polar proteins (MICROLON® 600) or medium binding surfaces, which are more hydrophobic and are therefore better suited for non-polar proteins and peptides (MICROLON® 200).
- Available with a choice of well profile: U-bottom, V-bottom, F-bottom (standard), F-bottom (chimney well), or half area
- Free of DNase, RNase and human DNA, and are non-pyrogenic
Specifikace
- Popis:96-well microplate, F-bottom (standard), MICROLON® 200, individually packed
- Sekvence:04
- Pracovní objem:25 - 340 µl
- Celkový objem:382 µl
- Cat. No.:GREI655001-000
- Certifikát ochrany životního prostředí:ISO 14001 (241002); ISO 50001 (241002)
- Environmentally Preferable:Low Manufacturing Impact