Orders LinkContactUsLinkComponent
Counting chambers
Counting chambers
Slides with ten chambers and large viewing area, without grid, 3×3 mm
Katalogové číslo 630-1539
Dodavatel:   Kova International
Counting chambers
Katalogové číslo 630-1539
Dodavatel:  Kova International
Counting chambers
Slides with ten chambers and large viewing area, without grid, 3×3 mm
Přejít na:
Price
4 860,00 Kč
Přihlásit se pro dostupnost
1 * 100 KS
Product Details & Documents
Designed to reduces pipetting errors and eliminate variable that could interfere with microscopic urinalysis (UA).
  • High optical quality through the composition acrylate
  • Perimeter contained wells for precise volume control and protection of cross-contamination and leakage
  • Capillary action for reproducible precision
  • Homogeneous suspension of sample sediments: Retained volume of 1 µl
  • For grid models, the grid is made up of nine squares, each divided into nine smaller squares
Grid: 3×3 mm
Volume retained on the grid: 1 µl
Specifikace
  • Type:
    without grid
  • Š×H:
    3×3 mm
  • Popis:
    Slides with ten chambers and large viewing area