SALT REGENERATION TABS KLOEK 1 * 25 kg
Katalogové číslo KING010235
Dodavatel: KING DISPOSABLES
Frequently Bought Together
Peroxid vodíku 30% unstabilised, AnalaR NORMAPUR® analytical reagent
Dodavatel: VWR Chemicals
Voda, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
Chlornan sodný 6…14% (active chlorine) in aqueous solution, GPR RECTAPUR®
Dodavatel: VWR Chemicals
Hypodermic needles, HENKE-JECT® for single use
Dodavatel: HENKE SASS WOLF
VWR®, Nitrile Gloves
Dodavatel: VWR Collection
VWR®, Ubrousky čisticí
Dodavatel: VWR Collection
Jodid draselný ≥99.5%, AnalaR NORMAPUR® analytical reagent
Dodavatel: VWR Chemicals
n-Heptan ≥99.0%, AnalaR NORMAPUR® analytical reagent
Dodavatel: VWR Chemicals
Barrels with lid and clamping ring
Dodavatel: RIXIUS
Chlorid sodný, EMSURE® ACS, ISO, Reag. Ph. Eur. p.a., Supelco®
Dodavatel: Merck
Filtry ze skleněných mikrovláken, grade GMF 150, Whatman™
Dodavatel: Whatman products (Cytiva)
Cover glasses, square
Dodavatel: GLASWARENFABRIK KARL HECHT
CPC® Quick-Disconnect Fittings, Push-to-Connect Coupling Bodies
Dodavatel: Avantor Fluid Handling
Bottle top vacuum filtration systems, Corning®
Dodavatel: Corning
Human plasma
Dodavatel: Biowest