Flame photometers, 360
01, Flame photometer, 360 industrial, 360 Industrial, (Na/K) 0,1 - 10 ppm (Li) 0,1 - 20 ppm (Ca) 2 - 400 ppm (Ba) 20 - 6000 ppm - Serum (only)
Katalogové číslo SHEW36000009
Dodavatel: SHERWOOD SCIENTIFIC INSTRUMENT
Přejít na:
Product Details & Documents
Single channel flame photometer for analysis of five elements (sodium, potassium, lithium, calcium, barium) designed with front loading, unobstructed sample work area and removable tray for easy cleaning.
- Automatic ignition and flame optimisation
- Safety features including fail safe flame detection and shut off with low air pressure indicator
- High sensitivity with fine manual control of air and stable flame design
- Indicator on the front panel indicate the filter selected and the flame status
- Useful display 'HOLD' button to ensure that readings are not lost before record
Delivery information: Supplied with Na, K, Ca, Ba and Li standard solutions (1000 ppm), reinforced nylon hose, tubing clip, drain tube, atomiser, nebuliser tubing and cleaning wire, PSU multi plug adapter and operating instructions.
Specifikace
- Model:360 Industrial
- Rozsah:(Na/K) 0,1 - 10 ppm (Li) 0,1 - 20 ppm (Ca) 2 - 400 ppm (Ba) 20 - 6000 ppm - Serum (only)
- Popis:Flame photometer, 360 industrial
- Sekvence:01
- Hmotnost:8,6 kg
- Cat. No.:SHEW36000009
- Š×H×V:420×350×480 mm
- Linearita:(Na/K/Li) ±2% at 3 ppm (Ca) ±2% at 100 ppm (Ba) ±2% at 1000 ppm
- Opakovatelnost:CV 1,0% for 20 aliquots sampled for 20 seconds using 10 ppm K solution
CV 1,5% for 20 consecutive samples using 10 ppm K solution
Accessories
Informace o produktu
- Katalogové čísloDostupnostPrice
- Nebuliser cleaning wires
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



