Storage Boxes
03, 24 compartments, 335×225×55 mm
Katalogové číslo SEMA5973
Dodavatel: SEMADENI
Přejít na:
Product Details & Documents
PP, translucent.
- Very stable
- Good chemical resistance
- Good temperature resistance
Multipurpose boxes for a wide range of uses.
Specifikace
- Popis:24 compartments
- Sekvence:03
- Š×H×V:335×225×55 mm