Orders LinkContactUsLinkComponent
Blocking buffers, FirstChoice™
Blocking buffers, FirstChoice™
03, FirstChoice™ blocking buffer-PBS, 500 mL
Katalogové číslo 786-668
Dodavatel:   G-Biosciences
Blocking buffers, FirstChoice™
Katalogové číslo 786-668
Dodavatel:  G-Biosciences
Číslo CAS:  
Blocking buffers, FirstChoice™
Blocking buffers, FirstChoice™
03, FirstChoice™ blocking buffer-PBS, 500 mL
Přejít na:
Číslo CAS:  
Price
3 750,00 Kč
Přihlásit se pro dostupnost
1 * 500 mL
Product Details & Documents
FirstChoice™ Blocking Buffer is ideal as a first choice for optimization of new assays and systems, or when determining the optimal blocking buffer for elimination of non specific binding sites in ELISA, blotting, immunohistochemistry and other applications. Buffers are conveniently supplied in widely used TBS (Tris-buffered saline at pH 7,5) and PBS (phosphate-buffered saline at pH 7,5) buffers as well as in separate formulations containing Tween® 20 for improving blocking efficiencies.
  • Ready to use
  • Greater versatility
  • Lack of cross reactivity
  • Compatible with antibodies and avidin/biotin based system
Specifikace
Specifikace
  • Bal.:
    500 mL
  • Popis:
    FirstChoice™ blocking buffer-PBS
  • Sekvence:
    03
Specifikace