Orders LinkContactUsLinkComponent
Threaded couplings
Threaded couplings
GL 32
Katalogové číslo BOHLH900-04
Dodavatel:   Bohlender
Threaded couplings
Katalogové číslo BOHLH900-04
Dodavatel:  Bohlender
Threaded couplings
Threaded couplings
GL 32
Přejít na:
Price
1 700,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Connector, screw thread coupling, made of PTFE. Two screw caps made of PPS or PBTP with GL thread and a PTFE/FPM gasket. The product is only exposed to PTFE.
  • Applicable for connecting two GL threads of different sizes
  • Temperature resisitance from −200 to +250 ºC
  • Very good chemical resistance
  • FDA conformity
Specifikace
  • Závit:
    GL 32