Orders LinkContactUsLinkComponent
Spore suspensions
Spore suspensions
01, Bacillus subtilis, cell line 6633: spore suspension (14 month shelf life), population 106/0,1 ml - steam sterilisation process
Katalogové číslo NAMSSBS-07
Dodavatel:   CROSSTEX
Spore suspensions
Katalogové číslo NAMSSBS-07
Dodavatel:  CROSSTEX
Spore suspensions
01, Bacillus subtilis, cell line 6633: spore suspension (14 month shelf life), population 106/0,1 ml - steam sterilisation process
Přejít na:
Price
25 670,00 Kč
Přihlásit se pro dostupnost
1 * 10 mL
Product Details & Documents
Documents
Specifikace
  • Popis:
    Bacillus subtilis, cell line 6633: spore suspension (14 month shelf life), population 106/0,1 ml - steam sterilisation process
  • Sekvence:
    01