Orders LinkContactUsLinkComponent
Boiling baths, SBB Aqua Plus series
Boiling baths, SBB Aqua Plus series
01, SBB Aqua 5 Plus, 100 °C, 1,3/1,5 kW, 5 L, 290×145×125 mm, 390×250×375 mm
Katalogové číslo 462-0171
Dodavatel:   GRANT INSTRUMENTS
Boiling baths, SBB Aqua Plus series
Katalogové číslo 462-0171
Dodavatel:  GRANT INSTRUMENTS
Boiling baths, SBB Aqua Plus series
Boiling baths, SBB Aqua Plus series
01, SBB Aqua 5 Plus, 100 °C, 1,3/1,5 kW, 5 L, 290×145×125 mm, 390×250×375 mm
Price
35 180,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Boiling baths are designed for continuous operation with water above 90 °C. They have a stainless steel tank in an outer case and analogue control by means of a simple dial for straightforward temperature settings. Baths have two fixed over-temperature safety cut-outs which can be reset by the user.
  • Energy regulator can be adjusted to ensure a steady boiling action, avoiding excessive steam or spitting
  • Constant level device maintains the required liquid level, avoiding the need to monitor and top up the bath
  • Heater and temperature sensors are mounted at the base of the tank, protected by a perforated tray, giving more usable space
  • Robust construction, durable and resistant to corrosion
  • New modern design
Required accessories must be ordered separately, see SUB Aqua Plus baths entry.
Informace pro objednávání: Required accessories must be ordered separately, see SUB Aqua Pro baths entry.
Informace o dodávce: Supplied with polycarbonate lid but without rings.
Upozornení: ** Required if flat-bottomed flasks are to be placed directly onto the base of the bath and to allow better convection in the bath.
Specifikace
  • Tepelný výkon:
    1,3/1,5 kW
  • Tepl.:
    100 °C
  • Kapacita:
    5 L
  • Sekvence:
    01
  • Š×H×V:
    390×250×375 mm
  • Š×H×V lázně:
    290×145×125 mm
  • Typ:
    SBB Aqua 5 Plus
Accessories
Informace o produktu
  • Katalogové číslo
    Sekvence
    Určeno pro
    Dostupnost
    Price
  • Stainless steel rack for 1.5 ml microtubes
    Stainless steel rack for 1.5 ml microtubes
    462-8458
    002
    65 tubes
    Přihlásit se pro dostupnost
    1 * 1 KS - 3 860,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Určeno pro
    Dostupnost
    Price
  • Stainless steel rack for 0.5 ml microtubes
    Stainless steel rack for 0.5 ml microtubes
    462-8457
    001
    105 tubes
    Přihlásit se pro dostupnost
    1 * 1 KS - 3 910,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Určeno pro
    Dostupnost
    Price
  • Stainless steel base tray**
    Stainless steel base tray**
    462-8350
    062
    SAP12, JBN12, JBA12, GLS Aqua 12 Plus , SBB Aqua 12 Plus
    Přihlásit se pro dostupnost
    1 * 1 KS - 2 970,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Určeno pro
    Dostupnost
    Price
  • Stainless steel base tray**
    Stainless steel base tray**
    462-8351
    063
    SAP18/26. JBN18/26, JBA18, GLS Aqua 18 Plus, OLS200 , SBB Aqua 18/26 Plus
    Přihlásit se pro dostupnost
    1 * 1 KS - 3 160,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Určeno pro
    Dostupnost
    Price
  • Stainless steel test tube rack for 30 mm Ø tubes
    Stainless steel test tube rack for 30 mm Ø tubes
    462-8456
    008
    12 tubes
    Přihlásit se pro dostupnost
    1 * 1 KS - 3 860,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Určeno pro
    Dostupnost
    Price
  • Stainless steel test tube rack for 19 mm Ø tubes
    Stainless steel test tube rack for 19 mm Ø tubes
    462-8454
    006
    32 tubes
    Přihlásit se pro dostupnost
    1 * 1 KS - 3 860,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Určeno pro
    Dostupnost
    Price
  • Stainless steel test tube rack for 25 mm Ø tubes
    Stainless steel test tube rack for 25 mm Ø tubes
    462-8455
    007
    18 tubes
    Přihlásit se pro dostupnost
    1 * 1 KS - 3 860,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Určeno pro
    Dostupnost
    Price
  • Stainless steel test tube rack for 16 mm Ø tubes
    Stainless steel test tube rack for 16 mm Ø tubes
    462-8453
    005
    36 tubes
    Přihlásit se pro dostupnost
    1 * 1 KS - 3 860,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Určeno pro
    Dostupnost
    Price
  • Stainless steel test tube rack for 13 mm Ø tubes
    Stainless steel test tube rack for 13 mm Ø tubes
    462-8452
    004
    55 tubes
    Přihlásit se pro dostupnost
    1 * 1 KS - 3 860,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Určeno pro
    Dostupnost
    Price
  • Stainless steel test tube rack for 10 mm Ø tubes
    Stainless steel test tube rack for 10 mm Ø tubes
    462-8451
    003
    84 tubes
    Přihlásit se pro dostupnost
    1 * 1 KS - 3 860,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Určeno pro
    Dostupnost
    Price
  • Gabled stainless steel lid
    Gabled stainless steel lid
    462-8212
    042
    JBA18, JBN18/26, SAP18/26, (SUB Aqua 18/26, JB Aqua 18/26, SBB Aqua 18/26, GLS Aqua 18 Plus), LSB18
    Přihlásit se pro dostupnost
    1 * 1 KS - 7 600,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Určeno pro
    Dostupnost
    Price
  • Gabled stainless steel lid
    Gabled stainless steel lid
    462-8211
    041
    JBA12, JBN12, SAP12, (SUB Aqua 12, SUB Aqua Dual, JB Aqua 12, JB Aqua Dual, SBB Aqua 12 Plus, GLS Aqua 12 Plus), LSB12
    Přihlásit se pro dostupnost
    1 * 1 KS - 6 770,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Určeno pro
    Dostupnost
    Price
  • Gabled stainless steel lid
    Gabled stainless steel lid
    462-8210
    040
    JBA5, JBN5, SAP2S, SAP5, (SUB Aqua 5, SUB Aqua Dual, JB Aqua 5, JB Aqua Dual, SBB Aqua 5 Plus)
    Přihlásit se pro dostupnost
    1 * 1 KS - 6 160,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Určeno pro
    Dostupnost
    Price
  • Flat lid with 6 ring sets
    Flat lid with 6 ring sets
    462-8222
    032
    JBA18, JBN18/26, SAP18/26, (SUB Aqua 18/26, JB Aqua 18/26, SBB Aqua 18/26 Plus)
    Přihlásit se pro dostupnost
    1 * 1 KS - 15 710,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Určeno pro
    Dostupnost
    Price
  • Flat lid with 4 ring sets
    Flat lid with 4 ring sets
    462-8221
    031
    JBA12, JBN12, SAP12, SAPD, (SUB Aqua 12, SUB Aqua Dual, JB Aqua 12, JB Aqua Dual, SBB Aqua 12 Plus)
    Přihlásit se pro dostupnost
    1 * 1 KS - 11 320,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Určeno pro
    Dostupnost
    Price
  • Raised half shelf with perforated mesh
    Raised half shelf with perforated mesh
    462-8232
    052
    JBN26, SAP26, (SUB Aqua 26, JB Aqua 26, SBB Aqua 26 Plus)
    Přihlásit se pro dostupnost
    1 * 1 KS - 6 560,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Určeno pro
    Dostupnost
    Price
  • Flat lid with 2 ring sets
    Flat lid with 2 ring sets
    462-8220
    030
    JBA5, JBN5, SAP5, SAPD, (SUB Aqua 5, SUB Aqua Dual, JB Aqua 5, JB Aqua Dual, SBB Aqua 5 Plus)
    Přihlásit se pro dostupnost
    1 * 1 KS - 7 360,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Určeno pro
    Dostupnost
    Price
  • Raised half shelf with perforated mesh
    Raised half shelf with perforated mesh
    462-0187
    051
    JBA18, JBN18, SAP18, (SUB Aqua 18, JB Aqua 18, SBB Aqua 18 Plus)
    Přihlásit se pro dostupnost
    1 * 1 KS - 5 520,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Určeno pro
    Dostupnost
    Price
  • Raised half shelf with perforated mesh
    Raised half shelf with perforated mesh
    462-8231
    050
    JBA12, JBN12, SAP12, SAPD, (SUB Aqua 12, SUB Aqua Dual, JB Aqua 12, JB Aqua Dual, SBB Aqua 12 Plus)
    Přihlásit se pro dostupnost
    1 * 1 KS - 4 690,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Určeno pro
    Dostupnost
    Price
  • Replacement lids, polycarbonate
    Replacement lids, polycarbonate
    GRANAQL5
    025
    SAP2S, SAP5 , JBN5, JBA5, SBB Aqua 5 Plus
    Přihlásit se pro dostupnost
    1 * 1 KS - 3 130,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Polycarbonate lid
    Polycarbonate lid
    462-0333
    03
    Přihlásit se pro dostupnost
    1 * 1 KS - 3 860,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Polycarbonate lid
    Polycarbonate lid
    462-0334
    01
    Přihlásit se pro dostupnost
    1 * 1 KS - 4 390,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Stainless steel base tray**, for SAP2S, SAP5, JBN5, JBA5 and SBB Aqua 5 Plus
    Stainless steel base tray**, for SAP2S, SAP5, JBN5, JBA5 and SBB Aqua 5 Plus
    462-8131
    061
    Přihlásit se pro dostupnost
    1 * 1 KS - 3 160,00 Kč
Informace o produktu
  • Katalogové číslo
    Materiál
    Ø
    Dostupnost
    Price
  • Anti-evaporation spheres
    Anti-evaporation spheres
    462-8239
    PP
    20 mm
    Přihlásit se pro dostupnost
    1 * 300 KS - 3 160,00 Kč