Orders LinkContactUsLinkComponent
Boiling baths, SBB Aqua Plus series
Boiling baths, SBB Aqua Plus series
04, SBB Aqua 26 Plus, 100 °C, 1,35/2,0 kW, 26 L, 290×145×175 mm, 390×600×445 mm
Catalog # 462-0170
Supplier:   GRANT INSTRUMENTS
Boiling baths, SBB Aqua Plus series
Catalog # 462-0170
Supplier:  GRANT INSTRUMENTS
Boiling baths, SBB Aqua Plus series
Boiling baths, SBB Aqua Plus series
04, SBB Aqua 26 Plus, 100 °C, 1,35/2,0 kW, 26 L, 290×145×175 mm, 390×600×445 mm
Price
47 490,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Boiling baths are designed for continuous operation with water above 90 °C. They have a stainless steel tank in an outer case and analogue control by means of a simple dial for straightforward temperature settings. Baths have two fixed over-temperature safety cut-outs which can be reset by the user.
  • Energy regulator can be adjusted to ensure a steady boiling action, avoiding excessive steam or spitting
  • Constant level device maintains the required liquid level, avoiding the need to monitor and top up the bath
  • Heater and temperature sensors are mounted at the base of the tank, protected by a perforated tray, giving more usable space
  • Robust construction, durable and resistant to corrosion
  • New modern design
Required accessories must be ordered separately, see SUB Aqua Plus baths entry.
Ordering information: Required accessories must be ordered separately, see SUB Aqua Pro baths entry.
Delivery information: Supplied with polycarbonate lid but without rings.
Caution: ** Required if flat-bottomed flasks are to be placed directly onto the base of the bath and to allow better convection in the bath.
Specifications
  • Tepelný výkon:
    1,35/2,0 kW
  • Tepl.:
    100 °C
  • Kapacita:
    26 L
  • Sekvence:
    04
  • Š×H×V:
    390×600×445 mm
  • Š×H×V lázně:
    290×145×175 mm
  • Typ:
    SBB Aqua 26 Plus
Accessories
Product Information
  • Catalog #
    Sekvence
    Popis
    Určeno pro
    Availability
    Price
  • 462-8458
    002
    Stainless steel rack for 1.5 ml microtubes
    65 tubes
    Log in for availability
    1 * 1 KS - 3 860,00 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Určeno pro
    Availability
    Price
  • 462-8457
    001
    Stainless steel rack for 0.5 ml microtubes
    105 tubes
    Log in for availability
    1 * 1 KS - 3 910,00 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Určeno pro
    Availability
    Price
  • 462-8350
    062
    Stainless steel base tray**
    SAP12, JBN12, JBA12, GLS Aqua 12 Plus , SBB Aqua 12 Plus
    Log in for availability
    1 * 1 KS - 2 970,00 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Určeno pro
    Availability
    Price
  • 462-8351
    063
    Stainless steel base tray**
    SAP18/26. JBN18/26, JBA18, GLS Aqua 18 Plus, OLS200 , SBB Aqua 18/26 Plus
    Log in for availability
    1 * 1 KS - 3 160,00 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Určeno pro
    Availability
    Price
  • 462-8456
    008
    Stainless steel test tube rack for 30 mm Ø tubes
    12 tubes
    Log in for availability
    1 * 1 KS - 3 860,00 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Určeno pro
    Availability
    Price
  • 462-8454
    006
    Stainless steel test tube rack for 19 mm Ø tubes
    32 tubes
    Log in for availability
    1 * 1 KS - 3 860,00 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Určeno pro
    Availability
    Price
  • 462-8455
    007
    Stainless steel test tube rack for 25 mm Ø tubes
    18 tubes
    Log in for availability
    1 * 1 KS - 3 860,00 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Určeno pro
    Availability
    Price
  • 462-8453
    005
    Stainless steel test tube rack for 16 mm Ø tubes
    36 tubes
    Log in for availability
    1 * 1 KS - 3 860,00 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Určeno pro
    Availability
    Price
  • 462-8452
    004
    Stainless steel test tube rack for 13 mm Ø tubes
    55 tubes
    Log in for availability
    1 * 1 KS - 3 860,00 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Určeno pro
    Availability
    Price
  • 462-8451
    003
    Stainless steel test tube rack for 10 mm Ø tubes
    84 tubes
    Log in for availability
    1 * 1 KS - 3 860,00 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Určeno pro
    Availability
    Price
  • 462-8212
    042
    Gabled stainless steel lid
    JBA18, JBN18/26, SAP18/26, (SUB Aqua 18/26, JB Aqua 18/26, SBB Aqua 18/26, GLS Aqua 18 Plus), LSB18
    Log in for availability
    1 * 1 KS - 7 600,00 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Určeno pro
    Availability
    Price
  • 462-8211
    041
    Gabled stainless steel lid
    JBA12, JBN12, SAP12, (SUB Aqua 12, SUB Aqua Dual, JB Aqua 12, JB Aqua Dual, SBB Aqua 12 Plus, GLS Aqua 12 Plus), LSB12
    Log in for availability
    1 * 1 KS - 6 770,00 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Určeno pro
    Availability
    Price
  • 462-8210
    040
    Gabled stainless steel lid
    JBA5, JBN5, SAP2S, SAP5, (SUB Aqua 5, SUB Aqua Dual, JB Aqua 5, JB Aqua Dual, SBB Aqua 5 Plus)
    Log in for availability
    1 * 1 KS - 6 160,00 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Určeno pro
    Availability
    Price
  • 462-8222
    032
    Flat lid with 6 ring sets
    JBA18, JBN18/26, SAP18/26, (SUB Aqua 18/26, JB Aqua 18/26, SBB Aqua 18/26 Plus)
    Log in for availability
    1 * 1 KS - 15 710,00 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Určeno pro
    Availability
    Price
  • 462-8221
    031
    Flat lid with 4 ring sets
    JBA12, JBN12, SAP12, SAPD, (SUB Aqua 12, SUB Aqua Dual, JB Aqua 12, JB Aqua Dual, SBB Aqua 12 Plus)
    Log in for availability
    1 * 1 KS - 11 320,00 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Určeno pro
    Availability
    Price
  • 462-8232
    052
    Raised half shelf with perforated mesh
    JBN26, SAP26, (SUB Aqua 26, JB Aqua 26, SBB Aqua 26 Plus)
    Log in for availability
    1 * 1 KS - 6 560,00 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Určeno pro
    Availability
    Price
  • 462-8220
    030
    Flat lid with 2 ring sets
    JBA5, JBN5, SAP5, SAPD, (SUB Aqua 5, SUB Aqua Dual, JB Aqua 5, JB Aqua Dual, SBB Aqua 5 Plus)
    Log in for availability
    1 * 1 KS - 7 360,00 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Určeno pro
    Availability
    Price
  • 462-0187
    051
    Raised half shelf with perforated mesh
    JBA18, JBN18, SAP18, (SUB Aqua 18, JB Aqua 18, SBB Aqua 18 Plus)
    Log in for availability
    1 * 1 KS - 5 520,00 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Určeno pro
    Availability
    Price
  • 462-8231
    050
    Raised half shelf with perforated mesh
    JBA12, JBN12, SAP12, SAPD, (SUB Aqua 12, SUB Aqua Dual, JB Aqua 12, JB Aqua Dual, SBB Aqua 12 Plus)
    Log in for availability
    1 * 1 KS - 4 690,00 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Určeno pro
    Availability
    Price
  • GRANAQL5
    025
    Replacement lids, polycarbonate
    SAP2S, SAP5 , JBN5, JBA5, SBB Aqua 5 Plus
    Log in for availability
    1 * 1 KS - 3 130,00 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Availability
    Price
  • 462-0333
    03
    Polycarbonate lid
    Log in for availability
    1 * 1 KS - 3 860,00 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Availability
    Price
  • 462-0334
    01
    Polycarbonate lid
    Log in for availability
    1 * 1 KS - 4 390,00 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Availability
    Price
  • 462-8131
    061
    Stainless steel base tray**, for SAP2S, SAP5, JBN5, JBA5 and SBB Aqua 5 Plus
    Log in for availability
    1 * 1 KS - 3 160,00 Kč
Product Information
  • Catalog #
    Popis
    Materiál
    Ø
    Availability
    Price
  • 462-8239
    Anti-evaporation spheres
    PP
    20 mm
    Log in for availability
    1 * 300 KS - 3 160,00 Kč